Diphenyl(1,5-cyclooctadiene)platinum(II)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
44039.03
44039.03EA
566
EUR
44039.03
Diphenyl(1,5-cyclooctadiene)platinum(II)
Diphenyl(1,5-cyclooctadiene)platinum(II)
CAS No.: 12277-88-2
Molecular Formula: C20H22Pt
Formula Weight: 457.47
Physical Form: Powder
Appearance: Colourless
MDL No.: MFCD03788255
Formel:
C₂₀H₂₂Pt Lagertemperatur: Raumtemperatur |
MDL:
MFCD03788255 CAS-Nummer: 12277-88-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...