2,3-Dimethoxyphenol ≥98%

Lieferant: Thermo Scientific
Achtung

44060.04
44060.04EA 59.6 EUR
44060.04
2,3-Dimethoxyphenol ≥98%
2,3-Dimethoxyphenol

Purity: 98% CAS No.: 5150-42-5 Molecular Formula: C8H10O3 Formula Weight: 154.16 Flash Point: 108°(226°F) Physical Form: Liquid Density: 1.182 Refractive Index: 1.540 Boiling Point: 233-234° MDL No.: MFCD00008366

Formel: C₈H₁₀O₃
Molecular Weight: 154,17 g/mol
Siedepunkt: 233…234 °C
Dichte: 1,182
Flammpunkt: 108 °C (226 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008366
CAS-Nummer: 5150-42-5
EINECS: 225-922-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR