Formamid 99+% Spectrophotometry

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Methanamid, Ameisensäureamid

44066.AE 44066.AP 44066.AY
44066.AEEA 50.7 EUR
44066.AE 44066.AP 44066.AY
Formamid 99+% Spectrophotometry
Formamid

Purity: 99+% CAS No.: 75-12-7 Molecular Formula: CH3NO Formula Weight: 45.04 Flash Point: 154°(310°F) Physical Form: Liquid Density: 1.134 Refractive Index: 1.4475 Sensitivity: Hygroscopic Appearance: Colourless Odour: Ammonia-like Melting Point: 2° Boiling Point: 210° Beilstein Reference: 505995 Merck Reference: 14,4237 MDL No.: MFCD00007941

Formel: HCONH₂
Molecular Weight: 45.04 g/mol
Siedepunkt: 210 °C
Schmelzpunkt: 2
Dichte: 1.134
Flammpunkt: 154 °C (310 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007941
CAS-Nummer: 75-12-7
EINECS: 200-842-0
Merck Index: 14,4237
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f381691a96baf5054ca8291e1fa9e9603b687e31&vid=e54dda1a985d7a4f9485c2cc5233498087e43f88&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR