Nickel plating solution, stromlos ammoniakfrei
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
44070.AK
44070.AKEA
29.2
EUR
44070.AK
Nickel plating solution, stromlos ammoniakfrei
Nickel plating solution
Appearance: Clear green
UN No.: UN3082
MDL No.: MFCD00011137
Applications: For plating on silicon, germainium, gallium arsenide, cadmium sulfide, nickel or Kovar
Lagertemperatur:
Raumtemperatur |
MDL:
MFCD00011137 UN: 3082 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...