Tetraammineplatinum(II) hydrogen phosphate-Lösung (0,5% Pt) (w/w)

Lieferant: Thermo Scientific
Gefahr

44076.NB
44076.NBEA 806 EUR
44076.NB
Tetraammineplatinum(II) hydrogen phosphate-Lösung (0,5% Pt) (w/w)
Tetraammineplatinum(II) hydrogen phosphate

CAS No.: 127733-98-6 Molecular Formula: H13N4O4PPt Formula Weight: 359.18 Physical Form: Liquid Density: 1.013 Appearance: Colourless MDL No.: MFCD03788258

Formel: H₁₃N₄O₄PPt
Molecular Weight: 359,18 g/mol
Dichte: 1,013
Lagertemperatur: Raumtemperatur
MDL: MFCD03788258
CAS-Nummer: 127733-98-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR