Poly(vinylidene fluoride)

Lieferant: Thermo Fisher Scientific

44080.36 44080.A3 44080.A9
44080.36EA 165 EUR
44080.36 44080.A3 44080.A9
Poly(vinylidene fluoride)
Poly(vinylidene fluoride)

Poly(vinylidene fluoride) is useful in coatings, film, filter cloth, instrument linings, filtration membranes, pump parts, linings and valves. It is also employed as a material for transducers in devices such as headphones, microphones and sonic detectors. It is also essential for piezoelectric and electrostrictive applications. In addition, it is useful for monofilament fishing lines replacing nylon monofilament. It acts as a standard binder used in the production of composite electrodes for lithium ion batteries. It serves as a high grade insulator for wires, nanomaterials and pressure sensors. It is utilized to make filters which are used to prepare samples for high performance liquid chromatography and other advanced analytical techniques.
Notes: Incompatible with strong oxidizing agents.


CAS No.: 24937-79-9 Molecular Formula: (-CH2CF2-)n Physical Form: Powder Appearance: White Odour: Odourless Melting Point: 155-160° MDL No.: MFCD00084470

Formel: (-CH₂CF₂-)n
Schmelzpunkt: 155…160 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00084470
CAS-Nummer: 24937-79-9

Bestellen

Spezifikation Testergebnisse

Appearance (Color) White
Form Powder
Viscosity Melt viscosity 23.50 to 29.50 Kpoise


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR