Bis(2-methoxyethyl)phthalat ≥96%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Bis(2-methoxyethyl) phthalate
44089.18
44089.18EA
34.9
EUR
44089.18
Bis(2-methoxyethyl)phthalat ≥96%
Bis(2-methoxyethyl)phthalat
Purity: 96%
CAS No.: 117-82-8
Molecular Formula: C14H18O6
Formula Weight: 282.29
Flash Point: 185°(365°F)
Physical Form: Liquid
Density: 1.17
Refractive Index: 1.503
Appearance: Colourless
Boiling Point: 340°
Beilstein Reference: 2056929
MDL No.: MFCD00042842
Formel:
C₁₄H₁₈O₆ Molecular Weight: 282,29 g/mol Siedepunkt: 340 °C Dichte: 1,17 Flammpunkt: 185 °C (365 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042842 CAS-Nummer: 117-82-8 EINECS: 204-212-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...