Stirring blades, moon-shaped
Lieferant: Bohlender
C400-06 C400-28 C400-26 C400-24 C400-12 C400-18 C400-22 C400-08 C400-14 C400-20 C400-16
441-9751EA 5.3 EUR
441-9751 BOHLC400-28 BOHLC400-26 BOHLC400-24 BOHLC400-12 BOHLC400-18 BOHLC400-22 BOHLC400-08 BOHLC400-14 BOHLC400-20 BOHLC400-16
Stirring blades, moon-shaped
Rührer Rührflügel
Stirrer blades made of PTFE, with double sided groove.
  • Resistant to chemicals
  • Temperature resistance from –200 to +250 °C
Suitable for glass stirrer shafts with double pivot, KPG glass stirrer shafts and PTFE-jacketed stainless steel stirrer shafts.