Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(II) dichloride
Lieferant: Thermo Fisher Scientific
44123.02
44123.04
44123.02EA
237
EUR
44123.02
44123.04
Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(II) dichloride
Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(II) dichloride
CAS No.: 36309-88-3
Molecular Formula: C72H48Cl2N6Ru
Formula Weight: 1169.20
Physical Form: Powder
MDL No.: MFCD03095387
Applications: Redox indicator, oxygen sensor
Formel:
C₇₂H₄₈Cl₂N₆Ru Lagertemperatur: Raumtemperatur |
MDL:
MFCD03095387 CAS-Nummer: 36309-88-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...