Gold ≥99,9% (Metall-Basis), dünne Folie, Dicke 0.002 mm (0.00008 in)
Lieferant: Thermo Fisher Scientific
Synonyme:
Aurum
44136.FF
44136.FI
44136.FFEA
522
EUR
44136.FF
44136.FI
Gold ≥99,9% (Metall-Basis), dünne Folie, Dicke 0.002 mm (0.00008 in)
Gold
Purity: 99.9%
CAS No.: 7440-57-5
Physical Form: ≈0.02g/25x25mm, not certified pinhole free
Odour: Odourless
MDL No.: MFCD00003436
Formel:
Au Molecular Weight: 196.97 g/mol Siedepunkt: 2800 Schmelzpunkt: 1063 Dichte: 19.3 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003436 CAS-Nummer: 7440-57-5 EINECS: 231-165-9 Merck Index: 12,04526 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...