Praseodym ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden), Pulver -40 mesh

Lieferant: Thermo Fisher Scientific
Gefahr


Unter Argon in Ampullen verpackt

44183.06
44183.06EA 210 EUR
44183.06
Praseodym ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden), Pulver -40 mesh
Praseodymium

Purity: 99.9% CAS No.: 7440-10-0 Physical Form: Ampouled under argon Sensitivity: Air and Moisture Sensitive Appearance: Grey to silver Odour: Odourless UN No.: 3089 MDL No.: MFCD00011174

Formel: Pr
Molecular Weight: 140.91 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011174
CAS-Nummer: 7440-10-0
EINECS: 231-120-3
UN: 3089
ADR: 4.1 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR