Silicium ≥99,9% (Metall-Basis), Pulver APS 1-5 µm

Lieferant: Thermo Fisher Scientific
Achtung

44185.09 44185.18 44185.30
44185.09EA 106 EUR
44185.09 44185.18 44185.30
Silicium ≥99,9% (Metall-Basis), Pulver APS 1-5 µm
Silicium

Purity: 99.9% CAS No.: 7440-21-3 Physical Form: Powder Appearance: White Odour: Odourless UN No.: 1346 MDL No.: MFCD00085311

Formel: Si
Lagertemperatur: Raumtemperatur
MDL: MFCD00085311
CAS-Nummer: 7440-21-3
EINECS: 231-130-8
UN: 1346
ADR: 4.1,III
Merck Index: 12,08635

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR