Organic reference standard, Isoamyl 4-methoxycinnamate, analytical standard, Supelco®
Lieferant: Merck
44189-50MG
44189-50MGEA 86.9 EUR
44189-50MG
Organic reference standard, Isoamyl 4-methoxycinnamate, analytical standard, Supelco®
Standards Organische Standards
Isoamyl 4-methoxycinnamate may be used as a reference standard in the determination of isoamyl 4-methoxycinnamate in water samples using liquid chromatography coupled with ultraviolet spectrophotometry detector (LC-UV).
  • Derivative of cinnamon acid ester
  • Utilised as a sunscreen agent
  • Act as a waterproof filter for ultraviolet-B (UV-B) rays
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ee88974fc380b3f7fb05b412225efe9cba5c704&vid=2ee88974fc380b3f7fb05b412225efe9cba5c704&prid=web&clid=SAPDAM