Organic reference standard, Isoamyl 4-methoxycinnamate, analytical standard, Supelco®
Lieferant: Merck
44189-50MG
44189-50MGEA
86.9
EUR
44189-50MG
Organic reference standard, Isoamyl 4-methoxycinnamate, analytical standard, Supelco®
Standards
Organische Standards
Isoamyl 4-methoxycinnamate may be used as a reference standard in the determination of isoamyl 4-methoxycinnamate in water samples using liquid chromatography coupled with ultraviolet spectrophotometry detector (LC-UV).
- Derivative of cinnamon acid ester
- Utilised as a sunscreen agent
- Act as a waterproof filter for ultraviolet-B (UV-B) rays