Bath, stainless steel, mixBATH
Lieferant: 2MAG
40950
442-0814EA 810 EUR
442-0814
Bath, stainless steel, mixBATH
Bäder Wasserbäder, ohne Umwälzung
Stainless steel mixBATH, durable, robust, is suitable for MIXdrive 6/15/60 as well as MIXdrive 1/1 eco/1 XS, fitting for immersion thermostats (heating/cooling), stirring point mark inside the bath, left-/right stop for optimised thermostat positioning (circulation pump) for use with MIXdrive 6 resp. MIXdrive 15.
  • Easy to clean bath
  • Even weld and electro-polished
  • Famed ridge formed as handle
  • Integrated drain makes emptying liquid easy and safe
  • Exact positioning of the stirrer so it is easy to use the stirrer under the bath