Magnetic stirring bars, PTFE, Spinfin®, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F371250014
442-9360EA 25.4 EUR
442-9360
Magnetic stirring bars, PTFE, Spinfin®, SP Bel-Art
Rührer Magnetrührstäbchen
Spinfin® magnetic stirring bars agitate liquids in round bottom vessels such as test tubes, cylinders and flasks where other stirring bars cannot be used. Bars maintain their position in a vessel and will not wobble.