Wolfram ≥99,95% (Metall-Basis), Pulver APS <1 µm

Lieferant: Thermo Fisher Scientific
Gefahr

44210.22 44210.36 44210.A3
44210.22EA 150 EUR
44210.22 44210.36 44210.A3
Wolfram ≥99,95% (Metall-Basis), Pulver APS <1 µm
Tungsten

Purity: 99.95% CAS No.: 7440-33-7 Appearance: Dark grey UN No.: 3089 MDL No.: MFCD00011461

Formel: W
Lagertemperatur: Raumtemperatur
MDL: MFCD00011461
CAS-Nummer: 7440-33-7
EINECS: 231-143-9
UN: 3089
ADR: 4.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR