Dodecyl 2-nitrophenyl ether ≥99,0%, Selectophore™ Ionophor, Supelco®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
Plasticizer for solvent polymeric membranes in ion-selective electrodes.
It was used to enhance extraction of nonpolar drugs during a study to demonstrate different approaches of basic drugs to maximize extraction recovery using electro membrane extraction method.
Formel:
C₁₈H₂₉NO₃ |
MDL:
MFCD00024221 CAS-Nummer: 83027-71-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...