Zinkbromid Hydrat ≥99,9% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr

44230.22 44230.36
44230.22EA 69.1 EUR
44230.22 44230.36
Zinkbromid Hydrat ≥99,9% (Metall-Basis)
Zinkbromid Hydrat

Purity: 99.9% Molecular Formula: ZnBr2.xH2O Formula Weight: 225.19(anhy) Physical Form: Crystalline Merck Reference: 14,10129 UN No.: 3260 MDL No.: MFCD00011294

Formel: Br₂Zn·H₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00011294
CAS-Nummer: 7699-45-8
EINECS: 231-718-4
UN: 3260
ADR: 8,II
Merck Index: 14,10129

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR