Organic reference standard, 3-Methylcholanthrene, analytical standard, Supelco®
Lieferant: Merck
442388
442388.EA 68.9 EUR
442388.
Organic reference standard, 3-Methylcholanthrene, analytical standard, Supelco®
Standards Organische Standards
3-Methylcholanthrene, a potent polycyclic aryl hydrocarbon receptor agonist, which is a carcinogen that can be obtained as a byproduct of combustion of various organic substances like coal, garbage, gas and oil.
It suppresses the proliferation of human umbilical vascular endothelial cells.