Dodecyltrimethylammoniumbromid, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

44239-10G
44239-10GEA 311 EUR
44239-10G
Dodecyltrimethylammoniumbromid, Supelco®
Dodecyltrimethylammonium bromide
Formel: C₁₅H₃₄BrN
Schmelzpunkt: 264…266 °C
MDL: MFCD00011767
CAS-Nummer: 1119-94-4
EINECS: 214-290-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=072ed8bdae3599bb34a6bee643dac22f63f4a306&vid=108f8d490db355eb500503d20580bda7bf3ae0ed&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR