n-Dotetracontan, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

44250-100MG
44250-100MGEA 93.8 EUR
44250-100MG
n-Dotetracontan, Supelco®
n-Dotetracontane
Formel: H₃C(CH₂)₄₀CH₃
Lagertemperatur: Raumtemperatur
CAS-Nummer: 7098-20-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=34865bc97fb11eed076e0fd7814ecdade558c06d&vid=5d44465d4e04ed99e3061af3405b7b0994e6743b&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR