Bis(pentafluorphenyl)phenylphosphin
Lieferant: Merck
Synonyme:
Bis(perfluorophenyl)(phenyl)phosphine, SpectraSynQ 443
442543
442543.EA
55.7
EUR
442543.
Bis(pentafluorphenyl)phenylphosphin
Bis(pentafluorphenyl)phenylphosphin
Formel:
C₁₈H₅F₁₀P Molecular Weight: 442,2 g/mol |
MDL:
MFCD00000291 CAS-Nummer: 5074-71-5 EINECS: 225-780-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...