trans-Chlordan

Lieferant: Merck

442599
442599.EA 255 EUR
442599.
trans-Chlordan
trans-Chlordane
Formel: C₁₀H₆Cl₈
CAS-Nummer: 5103-74-2
EINECS: 225-826-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75be2d17dab1124ee17090c7ed8164cb78b2dcfd&vid=50521a46362656f53531b916393e1c7dccca8265&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR