Magnesiumchlorid Hexahydrat 99,0 Complexometric, Molecular Biology Grade, Calbiochem®, Millipore®

Lieferant: Merck Millipore Calbiochem

Synonyme: Magnesium dichloride

442611-500
442611-500EA 85.7 EUR
442611-500
Magnesiumchlorid Hexahydrat 99,0 Complexometric, Molecular Biology Grade, Calbiochem®, Millipore®
Magnesiumchlorid Hexahydrat

  • DNases, proteases, RNases, none detected


Following reconstitution, store at room remperature (20°C). Stock solutions are stable for up to 6 months at room temperature.

Formel: MgCl₂·6H₂O
Molecular Weight: 203.3 g/mol
Siedepunkt: 1412
Schmelzpunkt: 117
Dichte: 1.57 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149781
CAS-Nummer: 7791-18-6
EINECS: 232-094-6
Merck Index: 13,05684

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR