Semi metals, plasma standard solution, As, Bi, Ga, Ge, In, Pb, Sb, Se, Sn, Te, Tl @ 100 µg/ml, Specpure®
Lieferant: Thermo Fisher Scientific
44268.AE
44268.AP
44268.AEEA
194
EUR
44268.AE
44268.AP
Semi metals, plasma standard solution, As, Bi, Ga, Ge, In, Pb, Sb, Se, Sn, Te, Tl @ 100 µg/ml, Specpure®
Standards
Kationischer Standards
Mehrelement-Standards
Molecular Formula: Matrix: 20% HCl/tr. HF
Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD00145276
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...