Semi metals, plasma standard solution, As, Bi, Ga, Ge, In, Pb, Sb, Se, Sn, Te, Tl @ 100 µg/ml, Specpure®

Lieferant: Thermo Fisher Scientific

44268.AE 44268.AP
44268.AEEA 194 EUR
44268.AE 44268.AP
Semi metals, plasma standard solution, As, Bi, Ga, Ge, In, Pb, Sb, Se, Sn, Te, Tl @ 100 µg/ml, Specpure®
Standards Kationischer Standards Mehrelement-Standards

Molecular Formula: Matrix: 20% HCl/tr. HF
Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD00145276

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR