Hochschmelzende Metalle, Plasma-Standardlösung, Al, B, Cr, Hf, Mo, Nb, Si, Ta, Ti, V, W, Zr @ 100 µg/ml, Specpure®
Lieferant: Thermo Fisher Scientific
44270.AE
44270.AP
44270.AEEA
239
EUR
44270.AE
44270.AP
Hochschmelzende Metalle, Plasma-Standardlösung, Al, B, Cr, Hf, Mo, Nb, Si, Ta, Ti, V, W, Zr @ 100 µg/ml, Specpure®
Standards
Kationischer Standards
Mehrelement-Standards
UN Nr.: UN3264
MDL Nr.: MFCD00151265
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...