Aqueous dispersant for multi-walled carbon nanotubes

Lieferant: Thermo Scientific
Achtung

44276.AB
44276.ABEA 64.9 EUR
44276.AB
Aqueous dispersant for multi-walled carbon nanotubes
Aqueous dispersant for multi-walled carbon nanotubes

Physical Form: Liquid MDL No.: MFCD04040667


Applications: For dispersing carbon nanotubes

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR