Tris(2-phenylpyridinato)iridium(III)
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Tris(2-phenylpyridine)iridium, Ir(ppy)3
44282.77
44282.77EA
220
EUR
44282.77
Tris(2-phenylpyridinato)iridium(III)
Tris(2-phenylpyridinato)iridium(III)
CAS No.: 693794-98-8
Molecular Formula: C33H24IrN3
Formula Weight: 654.82
Physical Form: Powder
Appearance: Yellow
Melting Point: 326-328°
MDL No.: MFCD04039766
Applications: As a green light emitting material
Formel:
C₃₃H₂₄IrN₃ Molecular Weight: 654,79 g/mol Schmelzpunkt: 326…328 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04039766 CAS-Nummer: 693794-98-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...