Ionic strength adjustor buffer solution for fluoride electrode, Supelco®
Lieferant: Merck
443123-475ML
443123-475MLEA 431 EUR
443123-475ML
Ionic strength adjustor buffer solution for fluoride electrode, Supelco®
Puffer Ionenstärkepuffer
It may be used as TISAB buffer in determination of fluoride in coal, coal fly ash, phosphate rock, limestone, mineral clay, fossilised materials, oyster tissue and vegetation using pyrohydrolysis for sample decomposition.