Blei(II)iodid, ultra trocken ≥99,999% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
Purity: 99.999%
CAS No.: 10101-63-0
Molecular Formula: I2Pb
Formula Weight: 461.00
Physical Form: -10 Mesh Beads
Density: 6.16
Appearance: Yellow
Odour: Odourless
Melting Point: 402°
Boiling Point: 954°
Merck Reference: 14,5411
UN No.: 2291
MDL No.: MFCD00011163
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...