Mangan(II)perchlorat Hexahydrat ≥99,995% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr

44318.09 44318.18
44318.09EA 91.7 EUR
44318.09 44318.18
Mangan(II)perchlorat Hexahydrat ≥99,995% (Metall-Basis)
Mangan(II)perchlorat Hexahydrat

Purity: 99.995% CAS No.: 15364-94-0 Molecular Formula: Mn(ClO4)2.6H2O Formula Weight: 361.93 (253.84anhy) Physical Form: Crystalline Sensitivity: Hygroscopic UN No.: 1481 MDL No.: MFCD00150255

Formel: Cl₂MnO₈·6H₂O
Schmelzpunkt: 165 °C, (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00150255
CAS-Nummer: 15364-94-0
EINECS: 237-390-9
UN: 1481
ADR: 5.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR