Chloroform, purified, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials
Gefahr

Synonyme: Trichlormethan

Filter 0,2 um

4432-28EA 0 EUR
4432-28 4432-10
Chloroform, purified, Macron Fine Chemicals™
Chloroform
Formel: CHCl₃
Molecular Weight: 119.38 g/mol
Siedepunkt: 61.7
Schmelzpunkt: –63 °C
Dichte: 1.4832 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00000826
CAS-Nummer: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1 III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use
Acid and Phosgene Passes Test
Aldehydes and Ketones (as HCHO) Passes Test
Chlorinated Decomposition Products and Chloride Passes Test
Free Chlorine Passes Test
Nonvolatile Residue (w/v) ≤0.002 %
Readily Carbonizable Substances Passes Test
Specific Gravity at 25°/25°C 1.476 - 1.480
Water (H₂O) Passes Test
Appearance (clear, colorless liquid)
Filtered through a 0.2 micron filter.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR