3-Methyl-1-phenyl-2-pyrazolin-5-on, Millipore®

Lieferant: Merck Millipore Calbiochem

Synonyme: MCI-186, 1-Phenyl-3-methyl-5-pyrazolone, 3-Methyl-1-phenyl-1H-pyrazol-5(4H)-one, Edaravone

443300-500
443300-500EA 114 EUR
443300-500
3-Methyl-1-phenyl-2-pyrazolin-5-on, Millipore®
3-Methyl-1-phenyl-2-pyrazolin-5-on
Formel: C₁₀H₁₀N₂O
Molecular Weight: 174,2 g/mol
MDL: MFCD00003138
CAS-Nummer: 89-25-8
EINECS: 201-891-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR