Aluminium ≥99,9999% (Metall-Basis), Drahtform, Puratronic®, geglüht, Ø 1.0 mm (0.04 in)
Lieferant: Thermo Fisher Scientific
44334.G5
44334.G9
44334.G5EA
197
EUR
44334.G5
44334.G9
Aluminium ≥99,9999% (Metall-Basis), Drahtform, Puratronic®, geglüht, Ø 1.0 mm (0.04 in)
Aluminium
Purity: 99.9999%
CAS No.: 7429-90-5
Physical Form: ≈2.1g/m, Temper: soft
Odour: Odourless
MDL No.: MFCD00134029
Formel:
Al Siedepunkt: 2327 °C (1013 hPa) Schmelzpunkt: 660 °C Dichte: 2,7 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00134029 CAS-Nummer: 7429-90-5 EINECS: 231-072-3 Merck Index: 13,00321 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...