Cobalt(II)oxid ≥99,995% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 99.995%
CAS No.: 1307-96-6
Molecular Formula: CoO, may contain up to 10% Co3O4
Formula Weight: 74.93
Physical Form: Powder
Density: 6.45
Appearance: Green-brown
Melting Point: 1935°
Merck Reference: 14,2446
UN No.: 2811
MDL No.: MFCD00016031
Formel:
CoO Schmelzpunkt: 1935 °C Dichte: 6,45 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016031 CAS-Nummer: 1307-96-6 EINECS: 215-154-6 UN: 2811 ADR: 6.1,III Merck Index: 14,2446 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...