Bis(cyclopentadienyl)zirkoniumdihydrid ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 98%
CAS No.: 37342-98-6
Molecular Formula: C10H12Zr
Formula Weight: 223.42
Physical Form: Powder
Sensitivity: Air and Moisture Sensitive
Appearance: White
UN No.: 3131
MDL No.: MFCD04126436
Hydrogenation catalyst for alkenes and alkynes
Formel:
C₁₀H₁₂Zr Lagertemperatur: Raumtemperatur |
MDL:
MFCD04126436 CAS-Nummer: 37342-98-6 UN: 3131 ADR: 4.3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...