N-Methyl-2-pyrrolidon (NMP) ≥99,0% ACS, Riedel-de Haën™

Lieferant: Honeywell Chemicals
Gefahr

443778-1L 443778-2.5L
443778-1LEA 162 EUR
443778-1L 443778-2.5L
N-Methyl-2-pyrrolidon (NMP) ≥99,0% ACS, Riedel-de Haën™
N-Methyl-2-pyrrolidon (NMP)

ACS GENERAL USE SOLVENTS


  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • Suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested to meet ACS general use specifications


Beilstein Number: 106420

Formel: C₅H₉NO
Siedepunkt: 204 °C (1,013 hPa)
Schmelzpunkt: –24 °C
Dichte: 1,028 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003193
CAS-Nummer: 872-50-4
EINECS: 212-828-1
Merck Index: 13,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32d156c242c81ef2974339109ced3895beda87fd&vid=06c87bbb3b5050362f47a8fc836cedc8a6d51530&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (GC) Min. 99.0 %
Water (Karl Fischer) Max. 0.05 %
Free amine Max. 0.01 %
Chloride (Cl) Max. 0.0001 %
APHA Max. 50
IR spectrum complying
Vapor Density 3.4 (vs air)
Vapor Pressure 0.99 mmHg ( 40 °C)
Autoignition Temperature 518 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR