Poly(styrene-divinylbenzene), aminomethylated, 1% quervernetzt mit Divinylbenzol 100-200 mesh
Lieferant: Thermo Fisher Scientific
44392.06
44392.14
44392.06EA
59.4
EUR
44392.06
44392.14
Poly(styrene-divinylbenzene), aminomethylated, 1% quervernetzt mit Divinylbenzol 100-200 mesh
Poly(styrene-divinylbenzene), aminomethylated
CAS No.: 89551-24-6
Physical Form: powder
MDL No.: MFCD00146442
Applications: Formation of amide groups using Boc strategy. As a scavenger resin
Lagertemperatur:
Raumtemperatur |
MDL:
MFCD00146442 CAS-Nummer: 89551-24-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...