Tris[N,N-bis(trimethylsilyl)amid]neodym(III) ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
44395.06
44395.06EA
442
EUR
44395.06
Tris[N,N-bis(trimethylsilyl)amid]neodym(III) ≥98%
Tris[N,N-bis(trimethylsilyl)amid]neodym(III)
Purity: 98%
CAS No.: 148274-47-9
Molecular Formula: C18H54N3NdSi6
Formula Weight: 625.39
Physical Form: Powder
Sensitivity: Moisture Sensitive
Appearance: Pale purple
Melting Point: 156-159°
UN No.: 3396
MDL No.: MFCD03427089
Formel:
C₁₈H₅₄N₃NdSi₆ Molecular Weight: 625,39 g/mol Schmelzpunkt: 156…159 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03427089 CAS-Nummer: 148274-47-9 UN: 3396 ADR: 4.3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...