VWR®, Support Rods
Lieferant: VWR Collection
444-0919EA 70 EUR
444-0919
VWR®, Support Rods
Stativstäbe
Heavy-duty, top-of-the-line rods provide maximum strength and durability. Constructed of 303 stainless steel.