Kreisförmig vibrierender Plattformschüttler, PSU-10i
Lieferant: GRANT INSTRUMENTS
PSU-10I
444-0933EA 978 EUR
444-0933
Kreisförmig vibrierender Plattformschüttler, PSU-10i
Schüttler Kreisschüttler
Orbital flatbed shaker, with LCD display of both set and actual speed, plus set and elapsed time. Available with a choice of four interchangeable accessory platforms, to enable use with a wide range of vessels, including bottles, flasks and beakers, dishes, boxes and petri dishes. An audible signal accompanies automatic switch-off to indicate the completion of a set time period. The soft start feature ensures a smooth ramp up of the shaking speed.
  • Variable speed control
  • Continuous or timer operation from 1 min to 96 h with automatic switch-off
  • Can be used at +4 to +40 °C, in an incubator or cold room
  • Low voltage power supply enables safe cold room operation
* Maximum speed is dependent on load.
Bestellinformation: Supplied without platform, required accessories must be ordered separately.