Shaking incubator, benchtop, Corning® LSE™
Lieferant: Corning
6791
444-1099EA 3785.88 EUR
444-1099
Shaking incubator, benchtop, Corning® LSE™
Schüttler Inkubationsschüttler
Truly a benchtop style, the lid on the benchtop shaking incubator has a low pivot point which provides a space-saving size sure to fit on the lab bench and under shelves.
Each unit includes a software package, which provides the option of saving data to an external computer or monitoring rpm and temperature throughout runs. The maximum capacity is 4×1 L flasks.