VWR®, Mini vortex mixer
Lieferant: VWR Collection
444-1337EA 152 EUR
444-1337
VWR®, Mini vortex mixer
Schüttler Vortex-Schüttler
The mini vortex mixer is the ideal vortex mixer for every laboratory with its compact size, ease of use, and its economic price point. The 3000 min⁻¹ fixed speed ensures there is no sacrificing any mixing power.
  • Extremely compact and easy to operate
  • Powerful vortexing for tubes up to Ø 30 mm
  • Refined steel base is designed to ensure product stability