VWR®, Tube rollers
Lieferant: VWR Collection
444-1339EA 1050 EUR
444-1339 444-1342
VWR®, Tube rollers
Schüttler Rotations-/Wippschüttler
Tube rollers are designed for the gentle, thorough mixing of a variety of containers, including Vacutainers™, bottles, test tubes, centrifuge tubes and roller bottles.
  • Variable speed between 2 and 80 min⁻¹
  • Timer from 1 minute to 100 hours, with an additional continuous mode
The VWR® tube rollers are designed for gentle yet complete mixing of a wide range of sample vessels, including Vacutainers™, media bottles, blood tubes, centrifuge tubes, and roller bottles. With both models, speed is controlled digitally.
In addition to the rolling motion, theMore Product Information