MultiPlate Genie® Microplate Shaker
Lieferant: SCIENTIFIC INDUSTRIES ELMIS
SI-4102
444-1430EA 721 EUR
444-1430
MultiPlate Genie® Microplate Shaker
Schüttler Mikrotiterplatten-Schüttler
Fully programmable microplate shaker for four plates (8 with optional tray).
  • Provides consistent well-to-well and plate-to-plate mixing for 96 and 384-well microplates, deep well plates, or PCR plates
  • Will accept PCR Work up racks with standard microplate footprint for use with PCR tubes and strips
  • Featuring programmable speed, time, pulsing/interval mixing - for accurate and repeatable results
Digital true vortex mixing microplate shaker handles up to 4 microplates, (or up to 8 plates with the optional accessory tray). The combined speed and 1 mm orbit provide consistant well to well and plate to plate uniformity, even in 384-well plates. Featuring programable speed, time and continuous or pulse time, it’s ideal for achieving accurate and repeatable results.
The Pulse feature allows rapid pulsing or timed ''on/off'' mixing for interval mixing. Ideal for use in PCR preparation, digestions, bead mixing, ELISA, EIA, and colorimetric assay applications.
Bestellinformation: Optional stacking tray accessory for additional 4 microplates (giving total of 8) is available.
Lieferumfang: Supplied with tray for 4 microplates.