Cole-Parmer® Stuart SI-200 Shaking Incubators
Lieferant: Cole Parmer
79520-04 79520-06 79520-16
444-1400EA 6850 EUR
444-1400 444-1433 444-1434
Cole-Parmer® Stuart SI-200 Shaking Incubators
Schüttler Mikrotiterplatten-Schüttler
Shaker and incubator combination makes these devices ideal for cell culture procedures.
  • Microprocessor controlled shaking speed ensures accuracy
  • Forced air circulation ensures uniform temperature inside the chamber
  • Easily set and view temperature and speed on the digital display
  • Unique retractable platform offers easy loading and unloading
These shaking incubators provide an orbital shaking motion to ensure even mixing while promoting bacterial growth. All models feature a timer which can be set from 1 second to 9 days. Once the timer has counted down, an alarm will sound and the shaking action will stop while the incubator will continue to run.
The retractable platform allows easy access to the samples at the back of the incubator yet securely locks in place during operation. A versatile clamping system holds flasks securely in place (up to 1 L flasks for 51 L models and up to 2 L flasks for 115 L models). Built-in USB connection and dedicated software allow long-term monitoring of the incubator temperature via a PC. Order models with IQ/OQ (Installation Qualification/Operational Qualification) documentation to help comply with regulatory agency requirements.
Temperature resolution: 0,1 °C
Temperature accuracy: ±0,5 °C
Lieferumfang: The 51 L models include USB cable, four securing bars, one UK (3-pin) power cord, and one EU (2-pin "Schuko") power cord. The 115 L model includes USB cable, four securing bars, two foot brackets, one UK (3-pin) power cord, and one EU (2-pin "Schuko") power cord.