Orbital vibrating platform, Bel-Art Spindrive®, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F370410001
444-2161EA 1170 EUR
444-2161
Orbital vibrating platform, Bel-Art Spindrive®, SP Bel-Art
Schüttler Kreisschüttler
Placed and centered on a magnetic stirrer, this platform operates as an orbital vibrator for plates, flasks or other vessels.
  • Economical
  • Compact design
  • Vibrate and mix; rotating orbit diameter of 1,52 mm (¹/₁₆")
  • Vibration adjustment is controlled by setting the magnetic stirrer to the desired speed
  • Base incorporates built-in hand holds for lifting and carrying
  • Supports up to 6,8 kg (15 lb)
Platform protects the operating mechanism from spills; non-skid surface to prevent containers from sliding.
Standards: U.S. Patent 5,409,312.
Bestellinformation: Supplied without magnetic stirrer which must be ordered separately.