Chloroform ≥99,8% (durch GC), AR® ACS, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials
Gefahr

Synonyme: Trichlormethan

4440-24EA 0 EUR
4440-24
Chloroform ≥99,8% (durch GC), AR® ACS, Macron Fine Chemicals™
Chloroform
Formel: CHCl₃
Molecular Weight: 119.38 g/mol
Siedepunkt: 61.7 °C (1013 hPa)
Schmelzpunkt: –63 °C
Dichte: 1.4832 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00000826
CAS-Nummer: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1 III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Meets ACS Reagent Chemical Requirements ≥99.8%
For Laboratory,Research,or Manufacturing Use Passes Test
Appearance (clear, colorless liquid) Passes Test
ACS - Assay (CHCl₃) (by GC) ≥99.8 %
ACS - Acetone, Aldehydes (as (CH₃)₂CO) Passes Test
ACS - Acid and Chloride Passes Test
ACS - Color (APHA) ≤10
ACS - Free Chlorine Passes Test
ACS - Lead (Pb) ≤0.050 ppm
ACS - Residue after Evaporation ≤0.001 %
ACS - Substances Darkened by H₂SO₄ Passes Test
ACS - Suitability for Use in Dithizone Tests Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR