Tris[N,N-bis(trimethylsilyl)amid]erbium(III) ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
unter Argon abgefüllt
44421.03
44421.06
44421.03EA
117
EUR
44421.03
44421.06
Tris[N,N-bis(trimethylsilyl)amid]erbium(III) ≥98%
Tris[N,N-bis(trimethylsilyl)amid]erbium(III)
Purity: 98%
CAS No.: 103457-72-3
Molecular Formula: C18H54ErN3Si6
Formula Weight: 648.41
Physical Form: Powder- Packaged under argon
Sensitivity: Moisture Sensitive
Appearance: Pink
Melting Point: 166°
UN No.: 3396
MDL No.: MFCD00271002
Formel:
C₁₈H₅₄ErN₃Si₆ Molecular Weight: 648.41 g/mol Schmelzpunkt: 166 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00271002 CAS-Nummer: 103457-72-3 UN: 3396 ADR: 4.3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...